Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFS8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104432
Description
Antibody detects endogenous levels of total NDUFS8.
Ndufs8 gene ontology annotations related to this gene include 4 iron, 4 sulfur cluster binding; oxidoreductase activity, acting on NAD(P)H.
Specifications
NDUFS8 | |
Polyclonal | |
Unconjugated | |
NDUFS8 | |
BC021616; CI-23k; CI23KD; CI-23kD; complex I 23kDa subunit; complex I-23kD; NADH dehydrogenase (ubiquinone) Fe-S protein 8; NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S8; NADH-ubiquinone oxidoreductase 23 kDa subunit; NDUFS8; TYKY; TYKY subunit | |
Rabbit | |
Affinity chromatography | |
RUO | |
225887, 293652, 4728 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O00217, Q8K3J1 | |
NDUFS8 | |
A synthesized peptide derived from human NDUFS8(Accession O00217), corresponding to amino acid residues L72-P122. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction