Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519344
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519344 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519344 Supplier Invitrogen™ Supplier No. PA519344
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine and human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

Ndufs8 gene ontology annotations related to this gene include 4 iron, 4 sulfur cluster binding; oxidoreductase activity, acting on NAD(P)H.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NDUFS8
Gene Accession No. O00217, Q8K3J1
Gene Alias BC021616; CI-23k; CI23KD; CI-23kD; complex I 23kDa subunit; complex I-23kD; NADH dehydrogenase (ubiquinone) Fe-S protein 8; NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S8; NADH-ubiquinone oxidoreductase 23 kDa subunit; NDUFS8; TYKY; TYKY subunit
Gene Symbols NDUFS8
Host Species Goat
Immunogen Synthetic peptide sequence (EAEIAANIQAD) corresponding to the internal amino acids of NDUFS8 (aa 196-206).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225887, 293652, 4728
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.