Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFV1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP133074 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP133074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP133074 Supplier Novus Biologicals Supplier No. NBP133074
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDUFV1 Polyclonal antibody specifically detects NDUFV1 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay.

Specifications

Antigen NDUFV1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Knockout Validated
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Alias CI-51K, CI-51kD, CI51KD, complex I 51kDa subunit, complex I, mitochondrial respiratory chain, Complex I-51kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, FLJ59059, mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1, NADH dehydrogenase (ubiquinone) flavoprotein 1 (51kD), NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa, NADH dehydrogenase flavoprotein 1, NADH-ubiquinone oxidoreductase 51 kDa subunit, UQOR1NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial
Gene Symbols NDUFV1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NDUFV1. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4723
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Zebrafish, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.