Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543156
Description
Peptide sequence: FMNKPSDGRP KYLVVNADEG EPGTCKDREI LRHDPHKLLE GCLVGGRAMG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.
Specifications
| NDUFV1 | |
| Polyclonal | |
| Unconjugated | |
| NDUFV1 | |
| CI-51K; CI51KD; CI-51kD; complex I 51 kda subunit; complex I 51kDa subunit; complex I, mitochondrial respiratory chain; complex I-51kD; mitochondrial NADH dehydrogenase (ubiquinone) 51kDa subunit; mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1; mitochondrial NADH:ubiquinone oxidoreductase 51 kda subunit; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) flavoprotein 1; NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa; NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial; NADH dehydrogenase flavoprotein 1; NADH dehydrogenase flavoprotein 1; 24 kDa (FP); NADH dehydrogenase ubiquinone flavoprotein 1; NADH:ubiquinone oxidoreductase core subunit V1; NADH-ubiquinone oxidoreductase 51 kDa subunit; NDUFV1; UQOR1; wu:fc01f01; wu:fc12f12; zgc:86620 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4723 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P49821 | |
| NDUFV1 | |
| Synthetic peptide directed towards the N-terminal of human NDUFV1 (aa 101-150). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction