Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFV1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985791
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5791 Supplier Bioss Supplier No. BS3959R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.

Specifications

Antigen NDUFV1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NDUFV1
Gene Accession No. P49821
Gene Alias BOS_24953; CI-51K; CI51KD; CI-51kD; complex I 51 kda subunit; complex I 51kDa subunit; complex I, mitochondrial respiratory chain; Complex I-51kD; mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1; mitochondrial NADH:ubiquinone oxidoreductase 51 kda subunit; NADH dehydrogenase (ubiquinone) flavoprotein 1; NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa; NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial; NADH dehydrogenase flavoprotein 1; NADH dehydrogenase flavoprotein 1; 24 kDa (FP); NADH dehydrogenase ubiquinone flavoprotein 1; NADH:ubiquinone oxidoreductase core subunit V1; NADH-ubiquinone oxidoreductase 51 kDa subunit; NDUFV1; UQOR1; wu:fc01f01; wu:fc12f12; zgc:86620
Gene Symbols NDUFV1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NDUFV1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4723
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.