Learn More
Invitrogen™ NDUFV3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total NDUFV3 protein.
Specifications
Specifications
| Antigen | NDUFV3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | NDUFV3 |
| Gene Accession No. | P56181 |
| Gene Alias | 1500032D16Rik; CI-10k; CI-9KD; complex I 10kDa subunit; complex I, mitochondrial respiratory chain, 10-kD subunit; complex I-9kD; Mipp65; mitochondrial NADH oxidoreductase-like protein; NADH dehydrogenase (ubiquinone) flavoprotein 3; NADH dehydrogenase (ubiquinone) flavoprotein 3, 10kDa; NADH dehydrogenase (ubiquinone) flavoprotein 3-like; NADH dehydrogenase [ubiquinone] flavoprotein 3, mitochondrial; NADH:ubiquinone oxidoreductase subunit V3; NADH-ubiquinone oxidoreductase 9 kD subunit; NADH-ubiquinone oxidoreductase 9 kDa subunit; NADH-ubiquinone oxidoreductase flavoprotein 3; NADH-ubiquinone oxidoreductase flavoprotein 3, 10kD; NDUFV3; Ndufv3l; renal carcinoma antigen NY-REN-4 |
| Gene Symbols | NDUFV3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.