Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NE 100 hydrochloride
GREENER_CHOICESDP

Catalog No. 313350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-335-0 50 mg
31-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-335-0 Supplier R&D Systems™ Supplier No. 3133/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective σ1 antagonist

Potent and selective σ1 receptor antagonist (Ki = 0.86 nM) that displays > 55-fold selectivity over σ2 receptors and > 6000-fold selectivity over D1, D2, 5-HT1A, 5-HT2 and PCP receptors. Exhibits reversible binding (Kd = 1.2 nM) and displays antipsychotic activity in vivo. Orally active.

Chemical Identifiers

CAS 149409-57-4
Molecular Formula C23H34ClNO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 391.97
Purity >98%
Molecular Weight (g/mol) 391.98
Chemical Name or Material NE 100 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.