Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NE-PER™ Nuclear and Cytoplasmic Extraction Reagents

Catalog No. pi78833
Change view
Click to view available options
Quantity:
15 mL
75 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78833 15 mL
PI78835 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI78833 Supplier Thermo Scientific™ Supplier No. 78833
Only null left
Add to Cart
Edge
Add to Cart

Efficiently lyse cells and extract separate cytoplasmic and nuclear protein fractions in less than two hours with this subcellular fractionation kit.

Thermo Scientific™ NE-PER Nuclear and Cytoplasmic Extraction Kit provides for efficient cell lysis and extraction of separate cytoplasmic and nuclear protein fractions in less than two hours.

The NE-PER Kit is a nuclear protein extraction method that involves simple, stepwise lysis of cells and centrifugal isolation of nuclear and cytoplasmic protein fractions. A benchtop microcentrifuge, tubes and pipettors are the only tools required. The NE-PER Reagents efficiently solubilize and separate cytoplasmic and nuclear proteins into fractions with minimal cross-contamination or interference from genomic DNA and mRNA. Once desalted or diluted, the isolated proteins can be used to perform immunoassays and protein interaction experiments, such as mobility shift assays (EMSA), co-immunoprecipitation (Co-IP) and pull-down assays.

Highlights:

  • Fast – obtain nuclear and cytoplasmic fractions in less than two hours
  • Proven – the NE-PER Kit is referenced in more than 950 distinct publications
  • Scalable – two kit sizes for producing extracts from cells and tissues
  • Convenient – simple instructions do not require ultracentrifugation over gradients
  • Compatible – use for downstream assays, including Western blotting, gel-shift assays, protein assays, reporter gene assays and enzyme activity assays

Includes:

Two kit sizes, each containing three buffer and detergent solutions

Compatible with:

Thermo Scientific LightShift Chemiluminescent EMSA Kit (Mfr. No. 20148)

Storage Requirements:

Upon receipt store kit components at 4°C

TRUSTED_SUSTAINABILITY

Specifications

Product Type Nuclear and Cytoplasmic Extraction Reagent
Content And Storage Upon receipt store kit components at 4°C.
Shipping Condition Ambient
Form Liquid
Product Line NE-PER
Buffer Lysis Buffer
Reagent Type Cell Lysis Buffer, Detergent Solution, Subcellular Fractionation
Quantity 15 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.