Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEBL Antibody, Novus Biologicals™
SDP

Catalog No. NBP145223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145223 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145223 Supplier Novus Biologicals Supplier No. NBP145223
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 4 publications

NEBL Polyclonal specifically detects NEBL in Human, Mouse, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen NEBL
Applications Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:10 - 1:500, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_998734.1
Gene Alias Actin-binding Z-disk protein, FLJ53769, LASP2, LIM-nebulette, LNEBL, MGC119746, MGC119747, nebulette
Gene Symbols NEBL
Host Species Goat
Immunogen Peptide with sequence C-ELQRLKRTQE corresponding to internal region according to NP_998734.1, NP_001166955.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10529
Test Specificity This antibody is expected to recognize isoform 2 and isoform 3 (NP_998734.1; NP_001166955.1) only, but does not recognize isoform 1 (NP_006384.1).
Target Species Human, Mouse, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.