Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEBL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553106 Supplier Invitrogen™ Supplier No. PA553106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQGIMNKEPA VIGRPDFEHA VEASKLSSQI KYKEKFDNEM KDKKHHYNPL ESASFRQNQL AATLASNVKY KKDIQNMHDP VSDLPNLLFL DHVLKASKML SGREYKKLFE ENKGMYHFDA DAVEHLHHKG NAVLQ Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

Nebulette encodes a nebulin like protein that is abundantly expressed in cardiac muscle. The encoded protein binds actin and interacts with thin filaments and Z-line associated proteins in striated muscle. This protein may be involved in cardiac myofibril assembly. A shorter isoform of this protein termed LIM nebulette is expressed in non-muscle cells and may function as a component of focal adhesion complexes. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEBL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NEBL
Gene Accession No. O76041
Gene Alias 1200007O21Rik; A630080F05Rik; Actin-binding Z-disk protein; BB140644; D830029A09Rik; LASP2; LIM and SH3 protein 2; LIM zinc-binding domain-containing Nebulette; LIM-nebulette; Lnebl; Nebl; Nebulette
Gene Symbols NEBL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEBL. Recombinant protein control fragment (Product #RP-90703).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10529
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.