Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nebulin Antibody (NB2), Novus Biologicals™
SDP

Catalog No. NB12011099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12011099 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12011099 Supplier Novus Biologicals Supplier No. NB12011099
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nebulin Monoclonal specifically detects Nebulin in Human, Mouse, Rat, Porcine, Alligator, Amphibian, Avian, Bovine, Chicken, Fish, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nebulin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone NB2
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:400
Formulation Ascites with 15mM Sodium Azide
Gene Alias DKFZp686C1456, FLJ11505, FLJ36536, FLJ39568, FLJ39584, NEB177D, nebulin, NEM2
Gene Symbols NEB
Host Species Mouse
Immunogen Full length native protein (purified) (Chicken).
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 4703
Test Specificity Localizes nebulin in skeletal muscle of a wide variety of species from cold-blooded vertebrates to man. Does not crossreact with smooth or cardiac muscle, non-muscle tissue and cultured cells. Localizes nebulin within the I band of skeletal muscle sections by indirect immunofluorescence.
Target Species Human, Mouse, Rat, Pig, Alligator, Amphibian, Avian, Bovine, Chicken, Fish, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.