Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nebulin Monoclonal Antibody (1F4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA519423
Description
Peptide Sequence: NTKTSYHTPA DMLSVTAAKD AQANITNTNY KHLIHKYILL PDAMNIELTR NMNRIQSDNE YKQDYNEWYK GLGWSPAGSL EVEKAKKATE YASDQKYRQH PSNFQFKKLT.
This gene encodes nebulin, a giant protein component of the cytoskeletal matrix that coexists with the thick and thin filaments within the sarcomeres of skeletal muscle. In most vertebrates, nebulin accounts for 3 to 4% of the total myofibrillar protein. The encoded protein contains approximately 30-amino acid long modules that can be classified into 7 types and other repeated modules. Protein isoform sizes vary from 600 to 800 kDa due to alternative splicing that is tissue-, species-, and developmental stage-specific. Of the 183 exons in the nebulin gene, at least 43 are alternatively spliced, although exons 143 and 144 are not found in the same transcript. Mutations in this gene are associated with recessive nemaline myopathy.
Specifications
Nebulin | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
P20929 | |
NEB | |
NEB (NP_004534, 2108 a.a. ~ 2217 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
1F4 | |
Unconjugated | |
NEB | |
AI595938; DKFZp686C1456; FLJ11505; FLJ36536; FLJ39568; FLJ39584; NEB; NEB177D; nebulin; NEM2; nemaline myopathy type 2 | |
Mouse | |
Affinity chromatography | |
RUO | |
4703 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction