Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nebulin Monoclonal Antibody (NB2)
GREENER_CHOICE

Catalog No. PIMA549308
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549308 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549308 Supplier Invitrogen™ Supplier No. MA549308
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: Mouse Skeletal Muscle Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes nebulin, a giant protein component of the cytoskeletal matrix that coexists with the thick and thin filaments within the sarcomeres of skeletal muscle. In most vertebrates, nebulin accounts for 3 to 4% of the total myofibrillar protein. The encoded protein contains approximately 30-amino acid long modules that can be classified into 7 types and other repeated modules. Protein isoform sizes vary from 600 to 800 kDa due to alternative splicing that is tissue-, species-, and developmental stage-specific. Of the 183 exons in the nebulin gene, at least 43 are alternatively spliced, although exons 143 and 144 are not found in the same transcript. Mutations in this gene are associated with recessive nemaline myopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nebulin
Applications Western Blot
Classification Monoclonal
Clone NB2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
Gene NEB
Gene Accession No. Q99439
Gene Alias AI595938; DKFZp686C1456; FLJ11505; FLJ36536; FLJ39568; FLJ39584; NEB; NEB177D; nebulin; NEM2; nemaline myopathy type 2
Gene Symbols NEB
Host Species Mouse
Immunogen Chicken breast nebulin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374027
Target Species Chicken
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.