Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NECAB1 Antibody (CL0576), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP25294625UL
Description
NECAB1 Monoclonal antibody specifically detects NECAB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NECAB1 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Mouse | |
Protein A purified | |
RUO | |
64168 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CL0576 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LLKETLNQLQSLQNSLECAMETTEEQTRQERQGPAKPEVLSIQWPGKRSSRRVQRHNSFSPNSP | |
25 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction