Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NECAB1 Antibody (OTI4G8), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272923AF350
Description
NECAB1 Monoclonal specifically detects NECAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
NECAB1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
NECAB1 | |
Full length recombinant protein of human NECAB1 (NP_071746) produced in HEK293T cells. | |
0.1 mL | |
64168.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI4G8 | |
Western Blot, Flow Cytometry | |
EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction