Learn More
Invitrogen™ NECAB1 Monoclonal Antibody (CL0580)

Description
Immunogen sequence: LLKETLNQLQ SLQNSLECAM ETTEEQTRQE RQGPAKPEVL SIQWPGKRSS RRVQRHNSFS PNSP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%. Binds to an epitope located within the peptide sequence SIQWPGKRSS as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | NECAB1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL0580 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NECAB1 |
| Gene Accession No. | Q8N987 |
| Gene Alias | 1700003H21Rik; EF hand calcium binding protein 1; EFCBP1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; Necab1; neuronal Ca(2+)-binding protein 1; neuronal calcium binding protein; neuronal calcium-binding protein 1; N-terminal EF-hand calcium binding protein 1; N-terminal EF-hand calcium-binding protein 1; Stip; Stip-1; synaptotagmin interacting protein 1; synaptotagmin-interacting protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.