Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NECAB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554849 Supplier Invitrogen™ Supplier No. PA554849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLKETLNQLQ SLQNSLECAM ETTEEQTRQE RQGPAKPEVL SIQWPGKRSS RRVQRHNSFS PNSP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

NECAB1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NECAB1
Gene Accession No. Q8N987
Gene Alias 1700003H21Rik; EF hand calcium binding protein 1; EFCBP1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; Necab1; neuronal Ca(2+)-binding protein 1; neuronal calcium binding protein; neuronal calcium-binding protein 1; N-terminal EF-hand calcium binding protein 1; N-terminal EF-hand calcium-binding protein 1; Stip; Stip-1; synaptotagmin interacting protein 1; synaptotagmin-interacting protein 1
Gene Symbols NECAB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NECAB1. Recombinant protein control fragment (Product #RP-93776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.