Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NECAB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555278
Description
Immunogen sequence: ASSQSGCLRI SIQKLSNESR YMIYEFWENS SVWNSHLQTN YSKTFQRSNV DFLETPELTS TMLVPASWW Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
NECAB1 is a protein coding gene.
Specifications
NECAB1 | |
Polyclonal | |
Unconjugated | |
NECAB1 | |
1700003H21Rik; EF hand calcium binding protein 1; EFCBP1; EF-hand calcium binding protein 1; EF-hand calcium-binding protein 1; Necab1; neuronal Ca(2+)-binding protein 1; neuronal calcium binding protein; neuronal calcium-binding protein 1; N-terminal EF-hand calcium binding protein 1; N-terminal EF-hand calcium-binding protein 1; Stip; Stip-1; synaptotagmin interacting protein 1; synaptotagmin-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64168 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N987 | |
NECAB1 | |
Recombinant protein corresponding to Human NECAB1. Recombinant protein control fragment (Product #RP-93778). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction