Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NECAB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553127 Supplier Invitrogen™ Supplier No. PA553127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATEDAKEEGL EAQISRLAEL IGRLESKALW FDLQQRLSDE DGTNMHLQLV RQEMAVCPEQ LSEFLDSLRQ YLRGTTGVRN CFHITAVRLS DGFTFVIYEF WETEEAWKRH LQSPLCKAFR HVKVDTLSQP EALSRILVPA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

The protein encoded by this gene is a neuronal calcium-binding protein that binds to and modulates the function of at least two receptors, adenosine A(2A) receptor and metabotropic glutamate receptor type 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAB2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NECAB2
Gene Accession No. Q7Z6G3
Gene Alias EF hand calcium binding protein 2; Efcbp2; EF-hand calcium binding protein 2; EF-hand calcium-binding protein 2; NECAB2; neuronal calcium binding 2; neuronal calcium-binding protein 2; N-terminal EF-hand calcium binding protein 2; N-terminal EF-hand calcium-binding protein 2; Stip-2; synaptotagmin-interacting protein 2
Gene Symbols NECAB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NECAB2. Recombinant protein control fragment (Product #RP-90838).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.