Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NECAB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584875 Supplier Invitrogen™ Supplier No. PA584875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGHLTDNLET EKLCDYFSEH LGVYRPVLAA LESLNRAVLA AMDATKLEYE R Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 90%.

The protein encoded by this gene interacts with the amino-terminal domain of the neuron-specific X11-like protein (X11L), inhibits the association of X11L with amyloid precursor protein through a non-competitive mechanism, and abolishes the suppression of beta-amyloid production by X11L. This protein, together with X11L, may play an important role in the regulatory system of amyloid precursor protein metabolism and beta-amyloid generation. The protein is phosphorylated by NIMA-related expressed kinase 2, and localizes to the Golgi apparatus. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NECAB3
Gene Accession No. Q96P71
Gene Alias 2900010M17Rik; AI853434; amyloid beta (A4) precursor protein-binding family A member 1 binding protein; amyloid beta (A4) precursor protein-binding, family A, member 1 binding protein; amyloid beta (A4) precursor protein-binding, family A, member 2 binding protein; amyloid beta A4 protein-binding family A member 2-binding protein; Amyloid-beta A4 protein-binding family A member 2-binding protein; Apba2bp; dJ63M2.4; dJ63M2.5; EFCBP3; EF-hand calcium binding protein 3; mXB51; NECAB3; Nek2-interacting protein 1; neuronal calcium-binding protein 3; neuronal calcium-binding protein NECAB3; NIP1; N-terminal EF-hand calcium binding protein 3; N-terminal EF-hand calcium-binding protein 3; STIP3; synaptotagmin interacting protein 2; synaptotagmin interacting protein STIP3; SYTIP2; X11L-binding protein 51; XB51
Gene Symbols NECAB3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NECAB3. Recombinant protein control fragment (Product #RP-108589).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63941
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.