Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NECAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | NECAP1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NECAP1 Polyclonal specifically detects NECAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NECAP1 | |
Polyclonal | |
Rabbit | |
Human | |
25977 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GQTIKLCIGNITNKKGGASKPRTA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
adaptin ear-binding coat-associated protein 1, adaptin-ear-binding coat-associated protein 1, DKFZP566B183, MGC131900, NECAP endocytosis associated 1, NECAP endocytosis-associated protein 1, NECAP-1 | |
NECAP1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title