Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NECAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100636
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100636 Supplier Invitrogen™ Supplier No. PA5100636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NECAP1.

Involved in endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Necap1
Gene Accession No. P69682, Q8NC96, Q9CR95
Gene Alias 1200016B17Rik; adaptin ear-binding clathrin-associated protein; adaptin ear-binding coat-associated protein 1; adaptin-ear-binding coat-associated protein 1; AI747569; brain enriched NECAP2 similar protein; clathrin-coated vesicle coat protein; EIEE21; hypothetical protein LOC504449; hypothetical protein MGC73124; mFLJ00061; NECAP endocytosis associated 1; NECAP endocytosis-associated protein 1; NECAP1; NECAP-1; RGD1306053; snoRNA MBI-129; zgc:73124
Gene Symbols Necap1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NECAP1(Accession Q8NC96), corresponding to amino acid residues E8-I58.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25977, 312694, 67602
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.