Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NECAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564106 Supplier Invitrogen™ Supplier No. PA564106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQTIKLCIGN ITNKKGGASK PRTA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 79%.

Involved in endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Necap1
Gene Accession No. Q8NC96
Gene Alias 1200016B17Rik; adaptin ear-binding clathrin-associated protein; adaptin ear-binding coat-associated protein 1; adaptin-ear-binding coat-associated protein 1; AI747569; brain enriched NECAP2 similar protein; clathrin-coated vesicle coat protein; EIEE21; hypothetical protein LOC504449; hypothetical protein MGC73124; mFLJ00061; NECAP endocytosis associated 1; NECAP endocytosis-associated protein 1; NECAP1; NECAP-1; RGD1306053; snoRNA MBI-129; zgc:73124
Gene Symbols Necap1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human NECAP1 (Product #RP-103502)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.