Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NECAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116577 Supplier Invitrogen™ Supplier No. PA5116577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse kidney, Mouse liver Immunogen sequence: GKTSTLIPPP GEQLAVGGSL VQPAVAPSSG GAPVPWPQPN PATADIWGDF TKSTGSTSSQ TQPGTGWVQF.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NECAP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Necap2
Gene Accession No. Q9D1J1
Gene Alias 1110005F07Rik; AA409105; adaptin ear-binding coat-associated protein 2; adaptin-ear-binding coat-associated protein 2; C78898; clathrin-coated vesicle coat protein; likely ortholog of H. sapiens adaptin-ear-binding coat-associated protein 2 (NECAP2); NECAP endocytosis associated 2; NECAP endocytosis-associated protein 2; Necap2; NECAP-2
Gene Symbols Necap2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 194-263 of human NECAP2 (NP_0605601).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66147
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.