Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Necdin Antibody (OTI5D1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272925AF405
Description
Necdin Monoclonal specifically detects Necdin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Necdin | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NDN | |
Full length recombinant protein of human Necdin (NP_002478) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5D1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
HsT16328, necdin, necdin (mouse) homolog, necdin homolog (mouse), PWCR | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Tumor Suppressors | |
4692.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction