Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Necdin Monoclonal Antibody (OTI5D1)
GREENER_CHOICE

Catalog No. PIMA526294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526294 Supplier Invitrogen™ Supplier No. MA526294
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Necdin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This intronless gene is located in the Prader-Willi syndrome deletion region. It is an imprinted gene and is expressed exclusively from the paternal allele.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Necdin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDN
Gene Accession No. Q99608
Gene Alias AI528698; HsT16328; Ndn; NECD; Necdin; necdin homolog; necdin, MAGE family member; necdin, melanoma antigen (MAGE) family member; necdin-like protein; Peg6; Prader-Willi syndrome chromosome region; PWCR
Gene Symbols NDN
Host Species Mouse
Immunogen Full length human recombinant protein of NDN produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4692
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.