Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Necdin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567347 Supplier Invitrogen™ Supplier No. PA567347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQLVQKAHELM WYVLVKDQKK MIIWFPDMVK DVIGSYKKWC RSILRRTSLI Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.

This intronless gene is located in the Prader-Willi syndrome deletion region. It is an imprinted gene and is expressed exclusively from the paternal allele.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Necdin
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDN
Gene Accession No. Q99608
Gene Alias AI528698; HsT16328; Ndn; NECD; Necdin; necdin homolog; necdin, MAGE family member; necdin, melanoma antigen (MAGE) family member; necdin-like protein; Peg6; Prader-Willi syndrome chromosome region; PWCR
Gene Symbols NDN
Host Species Rabbit
Immunogen Recombinant Human Necdin. Recombinant protein control fragment (Product #RP-107999).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.