Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Necdin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115635 Supplier Invitrogen™ Supplier No. PA5115635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Necdin.

This intronless gene is located in the Prader-Willi syndrome deletion region. It is an imprinted gene and is expressed exclusively from the paternal allele.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Necdin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NDN
Gene Accession No. P25233, Q99608
Gene Alias AI528698; HsT16328; Ndn; NECD; Necdin; necdin homolog; necdin, MAGE family member; necdin, melanoma antigen (MAGE) family member; necdin-like protein; Peg6; Prader-Willi syndrome chromosome region; PWCR
Gene Symbols NDN
Host Species Rabbit
Immunogen A synthesized peptide derived from human NDN(Accession Q99608), corresponding to amino acid residues A266-S316.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17984, 308690, 4692
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.