Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Necdin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129900
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129900 Supplier Invitrogen™ Supplier No. PA129900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat lysate..

This intronless gene is located in the Prader-Willi syndrome deletion region. It is an imprinted gene and is expressed exclusively from the paternal allele.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Necdin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 30% glycerol and 0.05% sodium azide; pH 7.4
Gene NDN
Gene Accession No. P25233, Q99608
Gene Alias AI528698; HsT16328; Ndn; NECD; Necdin; necdin homolog; necdin, MAGE family member; necdin, melanoma antigen (MAGE) family member; necdin-like protein; Peg6; Prader-Willi syndrome chromosome region; PWCR
Gene Symbols NDN
Host Species Rabbit
Immunogen Fusion protein, corresponding to N terminal amino acids 1-143 of Human Necdin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17984, 308690, 4692
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.