Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanSpace™ Technology CleanSpace™ Neck Pad

Thick foam neck pads are designed to be used with CleanSpace2 Respirators and can be attached to the unit using velcro strips.

Supplier:  CleanSpace™ Technology PAF0016

 View more versions of this product

Catalog No. 17-800-634

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

  • Used with the CleanSpace2 Respirators: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and offers breathability
  • Easy to wash and quick drying; quick to replace
  • Designed for long wear in harsh environments
  • Hygienic (medical quality materials) and hypoallergenic
Specifications

Specifications

AS/NZS1716: 2012, EN 12942
Foam
Thin
CleanSpace 2 Respirators
Store at -10°C to +55°C
8 g
Reusable
Comfort Pad
Welding, Woodworking, Manufacturing, Smelting, Construction, Mining, Agriculture, Grinding
PAF-0016
163 x 95 x 5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.