Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Necrostatin-1 Inhibitor
SDP

Catalog No. NB232410MG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232410MG 10 mg
NB232450MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB232410MG Supplier Novus Biologicals™ Supplier No. NB232410MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

ATP-competitive, allosteric inhibitor of receptor-interacting protein kinase 1 (RIPK1) (EC50 = 182nM)

  • ATP-competitive, allosteric inhibitor of receptor-interacting protein kinase 1 (RIPK1) (EC50 = 182 nM)
  • Blocks non-apoptotic cell death via inhibition of specific cellular pathway, necroptosis, which leads to necrosis (EC50 = 494nM)
  • Reduces ischemic brain injury in mouse model of stroke

Specifications

Purity >99%
Content And Storage Room Temperature
Quantity 10 mg
Product Type Necrostatin-1 Inhibitor
Molecular Weight (g/mol) 259.33

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.