Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nectin 1 Recombinant Rabbit Monoclonal Antibody (029)
GREENER_CHOICE

Catalog No. PIMA531079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531079 Supplier Invitrogen™ Supplier No. MA531079
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Rat CD111/Nectin-1/PVRL1.

This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nectin 1
Applications ELISA
Classification Recombinant Monoclonal
Clone 029
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Nectin1
Gene Accession No. 0
Gene Alias AI835281; AW549174; CD111; CLPED1; ectodermal dysplasia 4 (Margarita Island type); ED4; herpes simplex virus type 1 sensitivity; herpes virus entry mediator C; Herpesvirus entry mediator C; herpesvirus Ig-like receptor; HIgR; HV1S; Hvec; nectin 1; nectin cell adhesion molecule 1; nectin); Nectin1; nectin-1; nectin-1 alpha; nectin-1 delta; OFC7; poliovirus receptor-like 1; poliovirus receptor-related 1; poliovirus receptor-related 1 (herpesvirus entry mediator C; poliovirus receptor-related 1 (herpesvirus entry mediator C); poliovirus receptor-related 1 (herpesvirus entry mediator C; nectin); poliovirus receptor-related protein 1; PRR; PRR1; PVRL1; PVRR; PVRR1; SK-12
Gene Symbols Nectin1
Host Species Rabbit
Immunogen Recombinant Rat CD111/Nectin-1/PVRL1 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192183
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.