Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nectin-2/CD112 Antibody (110), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290530AF532
Description
Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Nectin-2/CD112 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CD112/ Nectin-2 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
110 | |
Western Blot, ELISA | |
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Nectin-2/CD112 extracellular domain (NP_033016.3; Met 1-Gly 351). | |
0.1 mL | |
Immunology | |
5819 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction