Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nectin-2/CD112 Antibody (610603), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2229PECY7
Description
Nectin-2/CD112 Monoclonal antibody specifically detects Nectin-2/CD112 in Human samples. It is validated for Flow CytometrySpecifications
Nectin-2/CD112 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
610603 | |
Flow Cytometry | |
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 | |
Mouse myeloma cell line NS0-derived recombinant human Nectin-2/CD112, Gln32-Gly360, Accession # Q92692 | |
0.1 mL | |
Immunology | |
5819 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction