Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin 2 Monoclonal Antibody (9D10G7E12), Invitrogen™

Mouse Monoclonal Antibody

$285.50 - $493.00

Specifications

Antigen Nectin 2
Clone 9D10G7E12
Concentration 1 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543525
View Documents
Invitrogen™
MA543525
50 μL
Each for $285.50
Only null left
Add to Cart
 
PIMA542963
View Documents
Invitrogen™
MA542963
100 μL
Each for $493.00
Only null left
Add to Cart
 
Description

Description

Specificity: Human CD112/Nectin2

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

Nectin 2
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative
AI325026; AI987993; CD112; herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; MPH; Murine herpes virus entry protein B; murine herpesvirus entry protein B; nectin cell adhesion molecule 2; Nectin2; Nectin-2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2; poliovirus sensitivity; PRR2; Pvr; Pvrl2; PVRR2; Pvs
NECTIN2
IgG1
Protein A
Antibody
9D10G7E12
ELISA
Unconjugated
Mouse
Human
Q92692
5819
Human full-length cell-derived rhCD112.
Primary
-20°C, Avoid Freeze/Thaw Cycles
NECTIN2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.