Learn More
Invitrogen™ Nectin 4 Polyclonal Antibody

Description
The antibody detects endogenous levels of total Nectin 4 protein.
Specifications
Specifications
| Antigen | Nectin 4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | NECTIN4 |
| Gene Accession No. | Q96NY8 |
| Gene Alias | 1200017F15Rik; EDSS1; ig superfamily receptor LNIR; LNIR; nectin 4; nectin cell adhesion molecule 4; NECTIN4; Nectin-4; poliovirus receptor-related 4; poliovirus receptor-related protein 4; Processed poliovirus receptor-related protein 4; PRR4; Pvrl4; RGD1559826 |
| Gene Symbols | NECTIN4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.