Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nectin 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598636
Description
Nectin 4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Nectin 4 (PVRL4) is a gene located on chromosome 1q23.3 that encodes a member of the nectin family of cellular adhesion molecules. These proteins are primarily involved in forming adherens junctions between epithelial cells, contributing to tissue integrity and maintenance in various tissues, including the skin, lungs, and mammary glands. Nectin 4 is highly expressed during embryonic development and is crucial for processes like tissue morphogenesis and repair. It acts as a ligand for the poliovirus receptor-related family and participates in cell signaling pathways that influence cell proliferation and survival. In the context of cancer, Nectin 4 is often upregulated, particularly in breast, ovarian, and lung cancers, where its expression is associated with tumor progression, metastasis, and poor prognosis. This makes Nectin 4 a potential target for therapeutic interventions and a candidate biomarker for cancer diagnosis and monitoring.
Specifications
Nectin 4 | |
Polyclonal | |
Unconjugated | |
NECTIN4 | |
1200017F15Rik; EDSS1; ig superfamily receptor LNIR; LNIR; nectin 4; nectin cell adhesion molecule 4; NECTIN4; Nectin-4; poliovirus receptor-related 4; poliovirus receptor-related protein 4; Processed poliovirus receptor-related protein 4; PRR4; Pvrl4; RGD1559826 | |
Rabbit | |
Protein G | |
RUO | |
81607 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.85 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96NY8 | |
NECTIN4 | |
Recombinant Human Nectin-4 protein (91-199AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction