Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nectin-4/PVRL4 Antibody (337516), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2659APCCY7
Description
Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for Flow CytometrySpecifications
Nectin-4/PVRL4 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
81607 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
337516 | |
Flow Cytometry | |
EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 | |
Mouse myeloma cell line NS0-derived recombinant human Nectin-4, Gly27-Val351, Accession # Q96NY8 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction