Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin 4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980634
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0634 Supplier Bioss Supplier No. BS10029R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Poliovirus receptor-like proteins (PVRLs), such as PVRL4, are adhesion receptors of the immunoglobulin superfamily and function in cell-cell adhesion.

Specifications

Antigen Nectin 4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NECTIN4
Gene Accession No. Q96NY8
Gene Alias 1200017F15Rik; EDSS1; Ig superfamily receptor LNIR; Lnir; nectin 4; Nectin cell adhesion molecule 4; Nectin4; Nectin-4; poliovirus receptor-related 4; Poliovirus receptor-related protein 4; Processed poliovirus receptor-related protein 4; Prr4; PVRL4; RGD1559826
Gene Symbols NECTIN4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Nectin 4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81607
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.