Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEDD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NEDD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NEDD1 Polyclonal specifically detects NEDD1 in Human samples. It is validated for Western Blot.Specifications
NEDD1 | |
Polyclonal | |
Rabbit | |
Q8NHV4 | |
121441 | |
IgG | |
72 kDa |
Western Blot | |
Unconjugated | |
RUO | |
FLJ35902, GCP-WD, NEDD-1, Neural precursor cell expressed developmentally down-regulated protein 1, neural precursor cell expressed, developmentally down-regulated 1, protein NEDD1, TUBGCP7 | |
NEDD1 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title