Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEDD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543970 Supplier Invitrogen™ Supplier No. PA543970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MQENLRFASS GDDIKIWDAS SMTLVDKFNP HTSPHGISSI CWSSNNNFLV Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEDD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Nedd1
Gene Accession No. Q8NHV4
Gene Alias GCP-WD; Nedd1; Nedd-1; NEDD1 gamma-tubulin ring complex targeting factor; neural precursor cell expressed developmentally down-regulated protein 1; neural precursor cell expressed, developmentally down-regulated 1; neural precursor cell expressed, developmentally down-regulated gene 1; protein NEDD1; TUBGCP7
Gene Symbols Nedd1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NEDD1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121441
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.