Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEDD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558455 Supplier Invitrogen™ Supplier No. PA558455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSTPNPKIAS SVTAGVASSL SEKIADSIGN NRQNAPLTSI QIRFIQNMIQ ETLDDFREAC HRDIVNLQVE MIKQFHMQLN EMHSLLERYS VNEGL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEDD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nedd1
Gene Accession No. Q8NHV4
Gene Alias GCP-WD; Nedd1; Nedd-1; NEDD1 gamma-tubulin ring complex targeting factor; neural precursor cell expressed developmentally down-regulated protein 1; neural precursor cell expressed, developmentally down-regulated 1; neural precursor cell expressed, developmentally down-regulated gene 1; protein NEDD1; TUBGCP7
Gene Symbols Nedd1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NEDD1. Recombinant protein control fragment (Product #RP-97798).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121441
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.