Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEDD1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990253
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0253 25 μL
50-199-0252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0253 Supplier Rockland Immunochemicals Supplier No. 109401C38S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with endogenous Nedd1 protein. A BLAST analysis was used to suggest reactivity with Nedd1 from human, chimpanzee, macaque, marmoset, cattle, rat, and mouse based on a 100% homology with the immunizing sequence. Expect partial reactivity with Nedd1 from turkey, chicken, salmon, and Danio based on a 91% homology with the immunizing sequence. Cross-reactivity with Nedd1 from other sources has not been determined.

Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.

Specifications

Antigen NEDD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 40 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Nedd1
Gene Accession No. Q8NHV4
Gene Alias BOS_5612; GCP-WD; Nedd1; Nedd-1; Neural precursor cell expressed developmentally down-regulated protein 1; neural precursor cell expressed, developmentally down-regulated 1; neural precursor cell expressed, developmentally down-regulated gene 1; Protein NEDD1; TUBGCP7
Gene Symbols Nedd1
Host Species Rabbit
Immunogen Anti-NEDD1 was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to the 343-667 region of human Nedd1.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121441
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.