Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat NEDD4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6218SP
This item is not returnable.
View return policy
Description
NEDD4 Monoclonal specifically detects NEDD4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NEDD4 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
NP_006145 | |
NEDD4 | |
E. coli-derived recombinant human NEDD4 Asn211-Gly349 (predicted) Accession # NP_006145 | |
25 μg | |
Primary | |
Detects human NEDD4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) NEDD8 or rhNEDD9 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
683211 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4734 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction