Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEDD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587453 Supplier Invitrogen™ Supplier No. PA587453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSYSSNGSDF GSCASITSGG SYTNSVISDS SSYTFPPSDD TFLGGNLPSD STSNRSVPNR NTTPCEIFSR STSTDPFVQD DLEHGLEIMK LPVSRNTKIP LKRYSSLVIF PRSPSTTRPT SPTSLCTLLS KGSYQTSHQF IISPSEIAHN EDGTSAKGFL STAVNGLRLS KTICTPGEVR DIRPLHRKGS LQKKIVLSNN TPRQTVCEKS SEGYSCVSVH FTQRKAATLD CETTNGDCKP EMSEIKLNSD SEYIKLMHRT SACLPSSQNV DCQININGEL ERPHSQMNKN HGILRRSISL G; Positive Samples: U-251MG, HeLa, Jurkat, A-549, SKOV3; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein.

The E3 ubiquitin-protein ligase NEDD4 is a Ubl conjugation pathway protein. It contains one HECT (E6AP-type E3 ubiquitin-protein ligase) domain and four WW domains. It is involved in the pathway leading to the degradation of VEGFR-2/KDFR, independently of its ubiquitin-ligase activity. It acts as a potential proto-oncogene that negatively regulates PTEN via ubiquitination, a paradigm analogous to that of Mdm2 and p53. It is also reported to be involved in the budding of many viruses and is the regulator of epithelial Na+ channels, which lead to arterial hypertension. NEDD4 protein is localized to the sarcolemmal region of muscle fibers. Increase in NEDD4 protein expression leads to unloaded and denervated muscles as well as a decrease in Notch1 expression. Hence it acts as down regulator of Notch-1 gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEDD4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NEDD4
Gene Accession No. P46934
Gene Alias AA959633; AL023035; AU019897; cell proliferation-inducing gene 53 protein; E3 ubiquitin-protein ligase NEDD4; E430025J12Rik; HECT-type E3 ubiquitin transferase NEDD4; Kiaa0093; mKIAA0093; NEDD4; NEDD-4; NEDD4 E3 ubiquitin protein ligase; Nedd4-1; Nedd4a; Neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated 4; neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4a; neural precursor cell expressed, developmentally down-regulted gene 4; OTTHUMP00000217840; OTTHUMP00000217841; OTTHUMP00000217842; PIG53; receptor-potentiating factor 1; RPF1
Gene Symbols NEDD4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-460 of human NEDD4 (NP_9406822).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4734
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.