Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEDD4 Rabbit anti-Human, Clone: 7R0M4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | NEDD4 |
---|---|
Clone | 7R0M4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NEDD4 Monoclonal antibody specifically detects NEDD4 in Human samples. It is validated for Western BlotSpecifications
NEDD4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4734 | |
IgG | |
Affinity purified |
7R0M4 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 | |
A synthetic peptide corresponding to a sequence within amino acids 1200-1300 of human NEDD4 (P46934). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title