Learn More
Invitrogen™ NEDD4L Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 22RV1 whole cell, human Jurkat whole cell, rat pancreas tissue, mouse pancreas tissue, mouse NIH3T3 whole cell. Flow: U20S cell.
Specifications
Specifications
| Antigen | NEDD4L |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | Antibody with 0.2mg Na2PO4, 0.9mg NaCl, 4mg trehalose and 0.05mg sodium azide |
| Gene | NEDD4L |
| Gene Accession No. | Q8CFI0, Q96PU5 |
| Gene Alias | 1300012C07Rik; E3 ubiquitin-protein ligase NEDD4-like; FLJ33870; HECT-type E3 ubiquitin transferase NED4L; hNEDD4-2; KIAA0439; NEDD4; NEDD4 like E3 ubiquitin protein ligase; NEDD4.2; Nedd4-2; nedd4b; NEDD4L; NEDL3; neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4b; neural precursor cell expressed, developmentally down-regulated gene 4-like; RSP5; ubiquitin-protein ligase Nedd4-2; ubiquitin-protein ligase Rsp5; Unknown (protein for IMAGE:7986262) |
| Gene Symbols | NEDD4L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.