Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEDD8 Antibody (5B8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$541.95
Specifications
| Antigen | NEDD8 |
|---|---|
| Clone | 5B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
NEDD8 Monoclonal specifically detects NEDD8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| NEDD8 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| FLJ43224, MGC104393, MGC125896, MGC125897, NEDD-8, Neddylin, Neural precursor cell expressed developmentally down-regulated protein 8, neural precursor cell expressed, developmentally down-regulated 8, Ubiquitin-like protein Nedd8 | |
| NEDD8 | |
| IgG1 | |
| Affinity Purified |
| 5B8 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cancer | |
| PBS with 0.05% Sodium Azide | |
| 4738 | |
| Purified recombinant fragment of human NEDD8 expressed in E. Coli. | |
| Primary | |
| 9 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title