Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NEDD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102372 Supplier Invitrogen™ Supplier No. PA5102372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NEDD8.

NEDD8 is a ubiquitin-like protein which plays an important role in cell cycle control and embryogenesis. Covalent attachment to its substrates requires prior activation by the E1 complex UBE1C-APPBP1 and linkage to the E2 enzyme UBE2M. Attachment of NEDD8 to cullins activates their associated E3 ubiquitin ligase activity, and thus promotes polyubiquitination and proteasomal degradation of cyclins and other regulatory proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEDD8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Nedd8
Gene Accession No. P29595, Q15843, Q71UE8
Gene Alias nedd8; NEDD-8; NEDD8 ubiquitin like modifier; nedd8.L; nedd8.S; neddylin; Neural precursor cell expressed developmentally down-regulated protein 8; neural precursor cell expressed, developmentally down-regulated 8; neural precursor cell expressed, developmentally down-regulated 8 L homeolog; neural precursor cell expressed, developmentally down-regulated 8 S homeolog; neural precursor cell expressed, developmentally down-regulated gene 8; neural precursor cell-expressed developmentally down-regulated; Rub1; si:zc14a17.5; ubiquitin; ubiquitin-like protein Nedd8; XELAEV_18007009mg
Gene Symbols Nedd8
Host Species Rabbit
Immunogen A synthesized peptide derived from human NEDD8(Accession Q15843), corresponding to amino acid residues M1-N51.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18002, 25490, 4738
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.