Learn More
Restek™ Needle Seat Assembly
SDP
Catalog No. 06611922 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric):
0.17 mm I.D.
Catalog No. Diameter (Metric)
06-611-922 0.17 mm I.D.
1 options
Catalog No. 06-611-922 Supplier Restek™ Supplier No. 25767
For Agilent HPLC systems
For Use With:
  • Agilent 1100
  • 1200
  • 1220
  • 1260
  • G1313A
  • G1392A/B
Specifications
Diameter (Metric) 0.17 mm I.D.
For Use With Agilent 1100, 1200, 1220, 1260, G1313A, G1392A/B
Volume (Metric) 2.3 μL
Product Type PEEK, Needle Seat